missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat CD25/IL-2R alpha Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB51561
This item is not returnable.
View return policy
Description
CD25/IL-2R alpha Monoclonal specifically detects CD25/IL-2R alpha in Rat samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| CD25/IL-2R alpha | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| P26897 | |
| IL2RA | |
| Chinese hamster ovary cell line CHO-derived recombinant rat CD25/IL-2 R alpha Glu22-Gln235 Accession # P26897 | |
| 100 μg | |
| Primary | |
| Detects rat CD25/IL-2 R alpha in ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) CD25/IL-2 R alpha, rmIL-2 R beta, rmCommon gamma chain, rmIL-15 R alpha, or recombinant human CD25/IL-2 R alpha is observed. | |
| Rat | |
| Purified |
| Flow Cytometry, CyTOF | |
| 745520 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD25, CD25 antigen, IDDM10, IL-2 R alpha, IL-2 receptor subunit alpha, IL2R, IL2R alpha, IL-2R subunit alpha, IL-2Ra, IL2RA, IL-2-RA, IL2-RA, interleukin 2 receptor, alpha, interleukin-2 receptor subunit alpha, p55, TAC antigen, TCGFR | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3559 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction