Learn More
Rat IgG VisUCyte HRP Polymer Antibody, R&D Systems™
GREENER_CHOICE
Product Code. 18776778
Change view
Click to view available options
Quantity:
125 mL
25 mL
250 mL (2 x 125 mL)
50 mL (2 x 25 mL)
Unit Size:
125mL
250mL
25mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18776778 125 mL 125mL
18714278 25 mL 25mL
18797818 50 mL (2 x 25 mL) 50mL
18766739 250 mL (2 x 125 mL) 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 18776778 Supplier R&D Systems Supplier No. VC005125
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Goat Polyclonal Antibody
IgG Polyclonal specifically detects IgG in Rat samples. It is validated for Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgG
Applications Immunohistochemistry
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry
Formulation Supplied as a solution in PBS with preservative.
Gene Alias Immunoglobulin G, ImmunoglobulinG
Host Species Goat
Immunogen Rat IgG
Purification Method Affinity Purified
Quantity 125 mL
Regulatory Status RUO
Primary or Secondary Secondary
Gene ID (Entrez) 3500
Target Species Rat
Content And Storage 6 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgG