missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rat IgG1 kappa Isotype Control (eBRG1), Brilliant Ultra Violet™ 805, eBioscience™, Invitrogen™
Rat Isotype Control
Brand: Affymetrix eBioscience 368-4301-81
Additional Details : Weight : 0.01000kg
Description
Rat IgG1 is a subclass of immunoglobulin G (IgG) antibodies that are commonly used in research and diagnostics. These antibodies are produced by rats in response to an antigen and can be purified from serum or hybridoma cell culture supernatants. Rat IgG1 is often used as a negative control in experiments, as it does not bind to many mammalian antigens. It is also used in immunohistochemistry, flow cytometry, and western blotting to detect specific antigens. Gene targeting is a powerful tool used to manipulate the rat genome and generate animal models for studying human diseases. With gene targeting, specific genes can be knocked out, inserted, or modified to investigate their role in disease pathogenesis.Specifications
Rat IgG1 kappa | |
Monoclonal | |
0.2 mg/mL | |
PBS with BSA and 0.09% sodium azide, pH 7.2 | |
Rat | |
50 μg | |
Isotype Control (Primary) | |
4°, C, store in dark, DO NOT FREEZE! | |
IgG1 κ |
Control, Flow Cytometry | |
eBRG1 | |
Brilliant Ultraviolet 805 | |
Immunoglobulin G, Immunoglobulin G1, ImmunoglobulinG, ImmunoglobulinG1 | |
Affinity chromatography | |
RUO | |
Not Applicable | |
Liquid |