Learn More
Novus Biologicals™ Rat Multi-tissue Tissue MicroArray (Normal)
Product Code. 18632841 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 Slides
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18632841 5 Slides Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18632841 Supplier Novus Biologicals™ Supplier No. NBP278074
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
The panel contains 5 to 8 different tissues on one slide.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 5 Slides
Species Rat
For Use With (Application) Please bake slides at 60C for 30 minutes before use.