Learn More
Novus Biologicals™ Rat Universal Protein Whole Tissue Lysate (Adult Whole Normal)
Shop All Bio Techne ProductsDescription
The Rat Universal Protein Whole Tissue Lysate (Adult Whole Normal) is derived from rat. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
| For Use With (Application) | Western Blot |
| Tissue | Universal |
| Quantity | 2 vials |
| Storage Buffer | HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors |
| Test Specificity | Universal Protein Lysate: Rat Normal Tissues |
| Cellular Fraction | Whole |
| Tissue Condition | Normal |
| Cross Reactivity | Rat |
| Host Species | Rat |
| Concentration | 5 mg/mL |
| Show More |