missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ravulizumab Recombinant Monoclonal Antibody
GREENER_CHOICE

Recombinant Monoclonal Antibody

477.00€ - 1204.00€

Specifications

Antigen Ravulizumab Humanized
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Classification Recombinant Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30283396
GREENER_CHOICE
View Documents
Invitrogen™
MA559285
100 μg
477.00€
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30283728
View Documents
Invitrogen™
MA559286
1 mg
1204.00€
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.

Ravulizumab is considered a long-acting complement 5 (C5) inhibitor that has been undergoing clinical trials for the treatment of paroxysmal nocturnal hemoglobinuria (PNH) as of 4 February, 2016. A pharmaceutical similar to ravulizumab (ALXN1210), called eculizumab, is currently approved for the treatment of PNH in 46 countries under the brand name Soliris™. Ravulizumab is considered by Alexion Pharmaceuticals Inc. to be a 'next-generation' eculizumab molecule. Ravulizumab was subsequently approved by the US FDA in December of 2018 for a variety of beneficial characteristics that make it an advanced, next-generation agent in comparison to eculizumab. In particular, ravulizumab is currently the first and only long-acting C5 complement inhibitor that can be administered every eight weeks for the treatment of adult patients with PNH whereas eculizumab is a bi-weekly treatment, Label. Moreover, virtually all of the phase 3 trial results for ravulizumab have demonstrated the equivalent efficacy and safety established by eculizumab and that patients transition safely and effectively from using eculizumab to ravulizumab. Subsequently, whereas PNH patients may have needed to previously plan their lives rather strictly around the bi-weekly infusion administrations of eculizumab, with ravulizumab such patients can find a more relaxed dosing schedule of only six or seven infusions over an entire year, Label.
Specifications

Specifications

Ravulizumab Humanized
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Lyophilized
IgG2SA
Human
ALXN-1210; ravulizumab-cwvz
Antibody
1 mg/mL
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
Unconjugated
Human
RUO
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.