missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Rb Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | Rb |
|---|---|
| Concentration | 0.14 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16887064
|
Proteintech
17218-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16877064
|
Proteintech
17218-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody is a rabbit polyclonal antibody raised against residues near the C terminus of human RB1.
ZNF639 binds DNA and may function as a transcriptional repressor.Specifications
| Rb | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P06400 | |
| 5925 | |
| Rb Fusion Protein Ag11035 | |
| Primary | |
| -20°C | |
| RB1 |
| 0.14 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| OSRC, p105 Rb, pp110, pRb, Rb, RB1, retinoblastoma 1 | |
| RB1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title