missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBCK1 Antibody (CL4290) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
422.00€ - 605.00€
Specifications
| Antigen | RBCK1 |
|---|---|
| Clone | CL4290 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279292
|
Novus Biologicals
NBP3-43640-100ul |
100 μL |
605.00€
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279740
|
Novus Biologicals
NBP3-43640-25ul |
25 μL |
422.00€
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RBCK1 Monoclonal antibody specifically detects RBCK1 in Human samples. It is validated for Western BlotSpecifications
| RBCK1 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Zinc Finger | |
| PBS, pH 7.2, 40% glycerol | |
| 10616 | |
| IgG1 | |
| Protein A purified |
| CL4290 | |
| Western Blot 1 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| C20orf18, EC 6.3.2.-, HBV associated factor 4, HBV-associated factor 4, Heme-oxidized IRP2 ubiquitin ligase 1, Hepatitis B virus X-associated protein 4, HOIL-1, RanBP-type and C3HC4-type zinc finger containing 1, ranBP-type and C3HC4-type zinc finger-containing protein 1, RBCC protein interacting with PKC1, RBCK2, RNF54RING finger protein 54, UBCE7IP3HOIL1, ubiquitin conjugating enzyme 7 interacting protein 3, Ubiquitin-conjugating enzyme 7-interacting protein 3, XAP3, XAP4chromosome 20 open reading frame 18, ZRANB4 | |
| This antibody was generated using a recombinant protein sequence of Q9BYM8, with the exact immunogen sequence remaining proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title