missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBCK1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26367-1-AP-20UL
This item is not returnable.
View return policy
Description
RBCK1 encodes a protein that is similar to mouse UIP28/UbcM4 interacting protein. Alternative splicing has been observed at this locus, resulting in distinct isoforms.Specifications
| RBCK1 | |
| Polyclonal | |
| Unconjugated | |
| RBCK1 | |
| C20orf18, HBV associated factor 4, HOIL 1, HOIL1, RBCK1, RBCK2, RING finger protein 54, RNF54, UBCE7IP3, XAP3, XAP4, ZRANB4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10616 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BYM8 | |
| RBCK1 | |
| RBCK1 Fusion Protein Ag24405 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction