missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM3 Antibody (CL0296) [FITC], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52906F
This item is not returnable.
View return policy
Description
RBM3 Monoclonal antibody specifically detects RBM3 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| RBM3 | |
| Monoclonal | |
| FITC | |
| IS1-RNPL, putative RNA-binding protein 3, RNA binding motif (RNP1, RRM) protein 3, RNA binding motif protein 3, RNA-binding motif protein 3, RNPL | |
| This antibody was developed using a recombinant protein derived from P98179, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| metabolism | |
| 5935 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0296 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction