Learn More
IKA RCT Basic Ready-to-go Solution
Product Code. 31941102 Shop All IKA Products
Change view
Click to view available options
Plug Type:
EU
Top Plate Material:
Aluminum Alloy
Ceramic-coated Aluminum
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type Top Plate Material Unit Size
31941102 EU Aluminum Alloy Each
31941089 EU Ceramic-coated Aluminum Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is under eco-contribution tax.
Product Code. 31941102 Supplier IKA Supplier No. 0010016459
  • ONLINE EXCLUSIVE PRICE
    970.00€
    889.73€ / Pack of 1
    Save 80.27€
    8% Off
View all promotions
 Restricted Item
Estimated Shipment: 18-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
Includes all components required for precise temperature control of your sample using a magnetic stirrer.
  • Stand for secure positioning
    • This complete solution includes a stand with an extension arm and a cross-sleeve
    • These ensure the correct and secure positioning of the PT 1000 temperature sensor, which is also supplied
    • This is because the sensor should always be positioned outside the vortex created during stirring and immersed at least 20 mm deep into the medium
    • Thanks to the stand, the position of the temperature sensor can be precisely adjusted to the reaction vessel and the liquid level in both vertical and horizontal directions
  • Multifunctional silicone cover
    • The package also includes a universal silicone cover for 600 mL and 1000 mL beakers
    • It features integrated openings for the temperature sensor
    • The chemically highly resistant cover fulfils three functions: it acts as a splash guard, minimises energy consumption when used as a lid, and reduces the noise that can be generated by air intake during rapid stirring
TRUSTED_SUSTAINABILITY
Specifications
Top Plate Material Aluminum Alloy
Voltage 220/230 V
Size 135 mm dia.
Temperature (Metric) 0°C to 310 °C
Type Magnetic Stirrer
Plug Type EU
No. of Positions 1
Max. Container Size 20 L
Stirring Range 50 to 1500 rpm
Warranty 2 Years
Certifications/Compliance CE
Display LED