missing translation for 'onlineSavingsMsg'
Learn More

IKA-Werke GmbH & Co. KG™ RCT Basic Synthesis Solution 250
GREENER_CHOICE

Product Code. 17785858 Shop All IKA Products
Change view
Click to view available options
Plug Type:
EU-Style
UK-Style
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plug Type unitSize
17785858 EU-Style Each
17795858 UK-Style Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.
Product Code. 17785858 Supplier IKA-Werke GmbH & Co. KG™ Supplier No. 0010011505
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under eco-contribution tax.

For safe synthesis in round-bottom flasks with 250 mL volume. The use of the synthesis blocks significantly reduces hazard potential compared to the use of oil baths and simplifies handling and cleaning of the round bottom flasks.

  • Universal clamp for secure mounting of synthesis flasks
  • Support rod for positioning temperature sensor
  • Elliptical magnetic stirring bars that perfectly adapt to the shape of the round-bottom flask
  • Stirring bar length: 20 to 80 mm
TRUSTED_SUSTAINABILITY

Specifications

Top Plate Material Aluminum Alloy
Voltage 220/230 V
Includes Support Rod, Universal Clamp, Stirring Bars (30 mm and 40 mm), 2 x Boss Head Clamps, 5 x IKAFLON 25 Ellipse, RCT Basic, PT 1000.6 Temperature Sensor, Holding Rod, Screw Driver, 250 mL Flask Carrier without Handle
Safety Features Temperature Safety Circuit
Shipping Weight (Metric) 2.4 kg
For Use With (Equipment) Connection For External Temperature Sensor PT1000, ETS-D5, ETS-D6
Height (Metric) 85 mm
Temperature (Metric) 310°C
Ambient Temperature 5°C to 40°C
Frequency 50/60 Hz
Width (Metric) 160 mm
Length (Metric) 270 mm
Wattage 650 W
Type Stirring Hotplate
Shape Round
Plug Type EU-Style
No. of Positions 1
Max. Container Size 20 L
Warranty 3 Year
Certifications/Compliance CE, TÜV (Standard IEC 61010-1 (cTÜVus))
Display LED
Top Plate Diameter (Metric) 135 mm
Speed 50 to 1500 rpm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.