Description
- Catalyzes the formation of a phosphodiester bond between the 5'-phosphoryl group and the 3'-hydroxyl group of two double-stranded DNA fragments
- Consists of a glassified reaction mix predispensed in single-reaction tubes
- When reconstituted to final reaction volume of 20μL, yields solution of reaction buffer, T4 DNA ligase, and ATP
Specifications
Specifications
| Components | FPLCpure™ T4 DNA Ligase 6 weiss units, 66mM Tris HCl (pH 7.6), 6.6mM MgCl2, 0.1mM ATP, 0.1mM Spermidine, 10mM DTT and stabilizer, glassified reaction mix predispensed in single-reaction tubes |
| For Use With (Application) | Self-circularization of linear DNA prior to transformation, ligation of insert to vector and ligation of synthetic linkers to blunt-ended DNA |
| Storage Buffer | TE Buffer (10mM Tris HCl, pH 8, with 1mM EDTA) or 1-2X One-phor-all Buffer |
| Content And Storage | Room Temperature |
| Quantity | 50 Reactions |
| Product Type | Ready-To-Go T4 DNA Ligase, 50 reaction beads |