Learn More
Cytiva Ready-To-Go™ T4 DNA Ligase
Product Code. 10238854 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
50 Reactions
Unit Size:
50 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10238854 50 Reactions 50 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10238854 Supplier Cytiva Supplier No. 27036101
For covalently linking DNA fragments in as little as 30 minutes
  • Catalyzes the formation of a phosphodiester bond between the 5'-phosphoryl group and the 3'-hydroxyl group of two double-stranded DNA fragments
  • Consists of a glassified reaction mix predispensed in single-reaction tubes
  • When reconstituted to final reaction volume of 20μL, yields solution of reaction buffer, T4 DNA ligase, and ATP
TRUSTED_SUSTAINABILITY
Specifications
Components FPLCpure™ T4 DNA Ligase 6 weiss units, 66mM Tris HCl (pH 7.6), 6.6mM MgCl2, 0.1mM ATP, 0.1mM Spermidine, 10mM DTT and stabilizer, glassified reaction mix predispensed in single-reaction tubes
For Use With (Application) Self-circularization of linear DNA prior to transformation, ligation of insert to vector and ligation of synthetic linkers to blunt-ended DNA
Storage Buffer TE Buffer (10mM Tris HCl, pH 8, with 1mM EDTA) or 1-2X One-phor-all Buffer
Content And Storage Room Temperature
Quantity 50 Reactions
Product Type Ready-To-Go T4 DNA Ligase, 50 reaction beads