Learn More
Thermo Electron LED GmbH Reagent Reservoir
GREENER_CHOICE
Product Code. 11781505
Change view
Click to view available options
Quantity:
10 Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11781505 10 Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11781505 Supplier Thermo Electron LED GmbH Supplier No. 9510037
  • ONLINE EXCLUSIVE PRICE
    40.73€
    36.41€ / Pack of 10
    Save 4.32€
    10% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The reagent reservoir from Thermo Electron LED GmbH is made of polypropylene and has eight wells.
  • Autoclavable
  • Provide variety of solutions to suit all pipetting procedures
  • Reagent reservoirs designed to provide solutions tailored to procedures
TRUSTED_SUSTAINABILITY
Specifications
Color White
Capacity (Metric) 60 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
Quantity 10 Pk.
Autoclavable Autoclavable
Channels 8
Disposable Reusable
Bottom Shape Flat