Learn More
Reagent Water, Lonza™
Product Code. 11650451
Change view
Click to view available options
Quantity:
30 mL
Unit Size:
30mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11650451 30 mL 30mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11650451 Supplier Lonza Supplier No. W50640
  • ONLINE EXCLUSIVE PRICE
    33.40€
    28.08€ / 30mL
    Save 5.32€
    15% Off
 Restricted Item
Estimated Shipment: 26-11-2026