Learn More
Heidolph™ Reax 2 with Universal Adapter
Product Code. 15674048 Shop All Heidolph Instruments Products
Change view
Click to view available options
Speed Range:
20 to 100 rpm
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Speed Range Unit Size
15674048 20 to 100 rpm 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: This product is under eco-contribution tax.
Product Code. 15674048 Supplier Heidolph™ Supplier No. 5412100900
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 09-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Regulatory: This product is under eco-contribution tax.
The Reax 2 with Universal Adapter quickly clamps in 2 vessels of any size and accepts load capacities up to 1 kg.
  • Speed continuously adjustable from 20 to 100 rpm
  • Universal adaptor allows for the use of various vessels of 50 to 160 mm height
  • In addition, an optional adaptor is available for the use of 20 test tubes at a time
TRUSTED_SUSTAINABILITY
Specifications
Load Bearing Capacity (Metric) 1 kg
Speed Range 20 to 100 rpm
Voltage 230 V
Product Type Shaker
Load Bearing Capacity (English) 2.2 lb.
Plug Type EU Plug
Model Reax 2