missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Chicken GH Antagonist Protein

A cDNA sequence encoding the GH Antagonist was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10641-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15934498

  • 125.00€ / 20µg

Estimated Shipment: 25-07-2023
to see stock.
Add to basket

Description

Description

Specifications

Specifications

GH Antagonist Protein
20 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
The protein was lyophilized from a concentrated (1 mg/ml) solution with 0.3% NaHCO3 pH 8.
Greater than 98.0% as determined by:(a) Analysis by SEC-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Chicken Growth Hormone G119R mutant did not bind to ovine GHR-ECD and was devoid of any biological activity in FDC-P1 3B9 cells. However, in binding experiments that were carried out using chicken liver membranes, both ovine GH and chicken GH showed similar IC50 values in competition with 125I-ovine GH, while the IC50 of G119R mutein was 10-fold higher. These results emphasize the importance of species specificity and indicate the possibility of antagonistic activity of chGH G119R in homologous system.
Chicken
Untagged
Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions