missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Cynomolgus Monkey CD58/LFA-3 Fc Protein, CF
Descrizione
Measured by its binding ability in a functional ELISA. Recombinant Cynomolgus Monkey CD58/LFA-3 Fc Chimera (Catalog # 11432-CD) binds to recombinant Human CD2 Fc Chimera Protein (Catalog # 1856-CD ) with a ED50 of 0.150-2.00 μg/mL.
Specifica
Specifica
| Accession Number | XP_0055542263.1 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Gene ID (Entrez) | 965 |
| Molecular Weight (g/mol) | M.W. (Observed): 70-78 kDa, under reducing conditions., M.W. (Theroretical): 48 kDa |
| Quantity | 50 μg |
| Source | Chinese Hamster Ovary cell line, CHO-derived cynomolgus monkey CD58/LFA-3 protein Cynomolgus Monkey CD58 (Ile29-Arg215) (N-terminus) Accession # XP_005542263.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU per 1 μg of the protein by the LAL method. |
| Gene Alias | Ag3, CD58 antigen, CD58 antigen, (lymphocyte function-associated antigen 3), CD58 molecule, FLJ23181, FLJ43722, LFA-3, LFA3ag3, lymphocyte function-associated antigen 3, Surface glycoprotein LFA-3 |
| Vedi altri risultati |
Titolo del prodotto
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.
Individuate un'opportunità di miglioramento?