missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Envelope-1 15kDa Protein
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Specifications
Specifications
| Name | Dengue Envelope-1 15kDa Protein |
| Quantity | 100 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Dengue Virus |
| Species | E. coli |
| Protein Tag | His |
| Buffer | 1x PBS pH 7.4. |
| Purity or Quality Grade | Protein is >95% pure as determined by 12% PAGE (coomassie staining). |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction