Learn More
enQuireBio™ Recombinant Dengue Virus Serotype 2 NS1 Protein
Recombinant Protein
Brand:  enQuireBio™ QP10478-500ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15923998
  • 1415.00€ / 500µg
Estimated Shipment: 28-06-2024
to see stock.
Description
Description
The NS1 non-structural protein, is an immunogenic protein, but poses no health risks and is therefore ideal for antibody production.
Specifications
Specifications
Serotype 2 NS1 Protein
Research Use Only
Recombinant Protein
Human
DPBS pH 7.4.
500 μg
Untested
Dengue virus (Strain: Thailand/16681/1984)
His
>95% as determined by SDS-PAGE.