missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Dengue Virus, ST4 Dengue NS1 Protein

Product Code. 15904058
Change view
Click to view available options
Quantity:
1 mg
100 μg
250 μg
500 μg
Unit Size:
100µg
1mg
250µg
500µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15904058 100 μg 100µg
15924058 250 μg 250µg
15934058 500 μg 500µg
15914058 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15904058 Supplier enQuireBio™ Supplier No. QP10497100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the Dengue NS1 was constructed and used to recombinantly synthesize the protein.

Specifications

Name Dengue Virus, ST4 Dengue NS1 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Untested
Product Type Recombinant Protein
Cross Reactivity Dengue Virus, ST4
Species Human
Protein Tag Native
Buffer 20mM Tris-HCl pH 7.5, 300mM NaCl.
Purity or Quality Grade >95% as determined by SDS-PAGE.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.