missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS5 Genotype-6 Protein
A cDNA sequence encoding the HCV NS5 Genotype-6 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12197-100ug
Additional Details : Weight : 0.01000kg
Specifications
Hepatitis C HCV NS5 Genotype-6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl, 1mM EDTA, 1.5M urea and 50% glycerol. |
100 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
Untagged | |
HCV NS5 Genotype-6 protein is >95% pure as determined by 10% PAGE (coomassie staining). |