missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant HIV-1 TAT Clade-D Protein

A cDNA sequence encoding the HIV-1 TAT Clade-D was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12265-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15909878

  • 1665.00€ / 100µg
Estimated Shipment: 01-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

HIV-1 TAT Clade-D Protein
Research Use Only
Recombinant Protein
E. coli
Lyophilized with 10% glycerol.
100 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
HIV
Untagged
Greater than 90.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions