missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human ADAM10 Protein

Product Code. 18137450
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18137450 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18137450 Supplier R&D Systems™ Supplier No. 936AD020

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

The Recombinant Human ADAM10 Protein is derived from Sf 21 (baculovirus). The Recombinant Human ADAM10 Protein has been validated for the following applications: Enzyme Activity.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Supplied as a 0.2 μm filtered solution in MES, NaCl, ZnCl 2Glycerol and Brij-35.
Gene ID (Entrez) 102
Molecular Weight (g/mol) 52 kDa
Name ADAM10
Quantity 20 μg
Source Spodoptera frugiperda,Sf 21 (baculovirus)-derived human ADAM10 protein Thr214-Glu672,with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70° C as supplied. 3 months, -70° C under sterile conditions after opening.
Structural Form Recombinant Human ADAM10 is prone to proteolytic cleavage at C-terminus. The predominant form of the purified protein lacks the His tag.
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 90%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.