missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL18 / PARC / MIP4 Protein
Recombinant Protein
Brand: enQuireBio™ QP5260-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002979.1 | |
CCL18 / PARC / MIP4 Protein | |
500 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human CCL18 / PARC / MIP4 induces primary human T cell chemotaxis. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
6362 | |
Greater than 95% as determined by SDS-PAGE. | |
Research Use Only | |
CCL18 | |
Recombinant Protein | |
E. coli | |
AQVGTNKELC CLVYTSWQIP QKFLVDYSET SPQCPKPGVI LLTKRGRQIC ADPNKKWVQK YISDLKLNA |