missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Complement Factor H-related 4/CFHR4

Product Code. 18767616
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18767616 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18767616 Supplier R&D Systems™ Supplier No. 5980CH050
 Restricted Item
Estimated Shipment: 04-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

His-tag

Measured by its binding ability in a functional ELISA. When Recombinant Mouse Complement Component C3d (Catalog # 2655-C3 ) is immobilized at 2 μg/mL (100 μL/well), Recombinant Human Complement Factor H-related 4/CFHR4 binds with an ED50 of 1-6 ng/mL.
TRUSTED_SUSTAINABILITY

Spezifikation

Accession Number CAA66980
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in Tris and NaCl with Trehalose.
Gene ID (Entrez) 10877
Molecular Weight (g/mol) M.W. (Observed): 39-45 kDa, reducing conditions; M.W. (theoretical): 37 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source Spodoptera frugiperda, Sf 21 (baculovirus)-derived human Complement Factor H-related 4/CFHR4 protein Gln19-Glu331, with a C-terminal 10-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, <= -20 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL aq. soln..
Buffer Lyophilized from a 0.2 μm filtered solution in Tris and NaCl with Trehalose
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Complement Factor H-related 4/CFHR4
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.