missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CRABP1 / RBP5 Protein

Product Code. 15918309
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15918309 1 mg 1mg
15908309 10 μg 10µg
15948309 50 μg 50µg
15998299 100 μg 100µg
15928309 200 μg 200µg
15938309 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15918309 Supplier enQuireBio™ Supplier No. QP5864ec1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) CRABP1 / RBP5, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_004369.1
Gene ID (Entrez) 1381
Name CRABP1 / RBP5 Protein
Quantity 1 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol CRABP1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence PNFAGTWKMR SSENFDELLK ALGVNAMLRK VAVAAASKPH VEIRQDGDQF YIKTSTTVRT TEINFKVGEG FEEETVDGRK CRSLPTWENE NKIHCTQTLL EGDGPKTYWT RELANDELIL TFGADDVVCT RIYVRE
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.