Learn More
enQuireBio™ Recombinant Human CRABP1 / RBP5 Protein
Product Code. 15998299
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15998299 100 μg 100µg
15908309 10 μg 10µg
15948309 50 μg 50µg
15928309 200 μg 200µg
15938309 500 μg 500µg
15918309 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 15998299 Supplier enQuireBio™ Supplier No. QP5864ec100ug
A DNA sequence encoding the Homo sapiens (Human) CRABP1 / RBP5, was expressed in the hosts and tags indicated.
Specifications
Accession Number NP_004369.1
Gene ID (Entrez) 1381
Name CRABP1 / RBP5 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol CRABP1
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag GST
Sequence PNFAGTWKMR SSENFDELLK ALGVNAMLRK VAVAAASKPH VEIRQDGDQF YIKTSTTVRT TEINFKVGEG FEEETVDGRK CRSLPTWENE NKIHCTQTLL EGDGPKTYWT RELANDELIL TFGADDVVCT RIYVRE
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.