missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human DEC-205/CD205 Fc Chimera Protein
Description
Measured by its binding ability in a functional ELISA. When Recombinant Human CD302/CLEC13A Fc Chimera (Catalog # 10203-CL ) is immobilized at 5 μg/mL (100 μL/well), Recombinant Human DEC-205/CD205 Fc Chimera (Catalog # 10205-DE) binds with an ED50 of 4-28 μg/mL.
Specifications
Specifications
| Accession Number | O60449.3 |
| For Use With (Application) | Bioactivity |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. |
| Molecular Weight (g/mol) | M.W. Observed: 200-250 kDa, under reducing conditions, M.W. Theoretical: 215 kDa |
| Quantity | 50 μg |
| Source | Human embryonic kidney cell, HEK293-derived human DEC-205/CD205 protein Human DEC-205/CD205 (Ser28-Asp1666) (N-terminus) Accession # O060449.3 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus) |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution. |
| Endotoxin Concentration | <0.10 EU / 1 μg of the protein by the LAL method. |
| Conjugate | Unconjugated |
| Reconstitution | Reconstitute at 500 μg/mL in PBS. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?