missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EIF4EBP3 Protein
A DNA sequence encoding the Homo sapiens (Human) EIF4EBP3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5974-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003723.1 | |
EIF4EBP3 Protein | |
Research Use Only | |
EIF4EBP3 | |
Recombinant Protein | |
E. coli | |
MSTSTSCPIP GGRDQLPDCY STTPGGTLYA TTPGGTRIIY DRKFLLECKN SPIARTPPCC LPQIPGVTTP PTAPLSKLEE LKEQETEEEI PDDAQFEMDI | |
Greater than 90% as determined by SDS-PAGE. |
8637 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |