missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Epiplakin Protein

A DNA sequence encoding the Homo sapiens (Human) Epiplakin, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5991-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929199

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

Specifications

Specifications

NP_112598.3
Epiplakin Protein
Research Use Only
EPPK1
Recombinant Protein
E. coli
MSGHTLPPLP VPGTNSTEQA SVPRAMAATL GAGTPPRPQA RSIAGVYVEA SGQAQSVYAA MEQGLLPAGL GQALLEAQAA TGGLVDLARG QLLPVSKALQ QGLVGLELKE KLLAAERATT GYPDPYGGEK LALFQAIGKE VVDRALGQSW LEVQLATGGL VDPAQGVLVA PEPACHQGLL DRETWHKLSE LEPGTGDLRF LNPNTLERLT YHQLLERCVR APGSG
Greater than 90% as determined by SDS-PAGE.
83481
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions