missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Epiplakin Protein
A DNA sequence encoding the Homo sapiens (Human) Epiplakin, was expressed in the hosts and tags indicated.
160.00€ - 2385.00€
Specifications
Accession Number | NP_112598.3 |
---|---|
Gene ID (Entrez) | 83481 |
Name | Epiplakin Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15979189
|
enQuireBio™
QP5991-EC-100UG |
100 μg |
530.00€
100µg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15999189
|
enQuireBio™
QP5991-EC-1MG |
1 mg |
2385.00€
1mg |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15989189
|
enQuireBio™
QP5991-EC-10UG |
10 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15909199
|
enQuireBio™
QP5991-EC-200UG |
200 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15919199
|
enQuireBio™
QP5991-EC-500UG |
500 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
15929199
|
enQuireBio™
QP5991-EC-50UG |
50 μg |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Specifications
NP_112598.3 | |
Epiplakin Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
83481 | |
Research Use Only | |
EPPK1 | |
Recombinant Protein | |
E. coli | |
MSGHTLPPLP VPGTNSTEQA SVPRAMAATL GAGTPPRPQA RSIAGVYVEA SGQAQSVYAA MEQGLLPAGL GQALLEAQAA TGGLVDLARG QLLPVSKALQ QGLVGLELKE KLLAAERATT GYPDPYGGEK LALFQAIGKE VVDRALGQSW LEVQLATGGL VDPAQGVLVA PEPACHQGLL DRETWHKLSE LEPGTGDLRF LNPNTLERLT YHQLLERCVR APGSG | |
Greater than 90% as determined by SDS-PAGE. |