missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human FABP1 Protein
A cDNA sequence encoding the FABP1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11830-10ug
Additional Details : Weight : 0.01000kg
Specifications
FABP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MSFSGKYQLQ SQENFEAFMK AIGLPEELIQ KGKDIKGVSE IVQNGKHFKF TITAGSKVIQ NEFTVGEECE LETMTGEKVK TVVQLEGDNK LVTTFKNIKS VTELNGDIIT NTMTLGDIVF KRISKRI | |
Greater than 90.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
Filtered (0.4 microm) and lyophilized from 0.5 mg/ml in 20mM TRIS and 50mM NaCl, pH 7.5. |