Learn More
R&D Systems™ Recombinant Human FCRN Protein
Product Code. 18718617
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18718617 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18718617 Supplier R&D Systems™ Supplier No. 8639FC050
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
Non-covalently associated with endogenous beta 2-Microglobulin
TRUSTED_SUSTAINABILITY
Specifications
Accession Number P55899
For Use With (Application) Bioactivity
Gene ID (Entrez) 2217
Molecular Weight (g/mol) Observed M.W.: 30-36kDa (FCRN), 10kDa ( beta 2 Microglobulin), Theoretical M.W.: 31kDa (FCRN), 10kDa ( beta 2 Microglobulin)kDa
Quantity 50 μg
Source Human embryonic kidney cell,HEK293-derived human FCRN protein Ala24-Ser297,with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Gene Alias alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FCGRT, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig
Common Name FCRN/FCGRT
Gene Symbol FCGRT
Biological Activity Measured by its binding ability in a functional ELISA. When Recombinant Human FCRN is immobilized at 2μg/mL (100μL/well), the concentration of biotinylated human IgG that produces 50% of the optimal binding response is approximately 2-10μg/mL.
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Expression System Human embryonic kidney cell, HEK293-derived Ala24-Ser297, with a C-terminal 6-His tag
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining