missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human GLTPD1 Protein
A cDNA sequence encoding the GLTPD1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP11994-10ug
Additional Details : Weight : 0.01000kg
Specifications
80772 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDDSETG FNLKVVLVSF KQCLDEKEEV LLDPYIASWK GLVRFLNSLG TIFSFISKDV VSKLRIMERL RGGPQSEHYR SLQAMVAHEL SNRLVDLERR SHHPESGCRT VLRLHRALHW LQLFLEGLRT SPEDARTSAL CADSYNASLA AYHPWVVRRA VTVAFCTLPT REVFLEAMNV GPPEQAVQML GEALPFIQRV YNVSQKLYAE HSLLDLP | |
Greater than 90.0% as determined by SDS-PAGE. |
GLTPD1 Protein | |
Research Use Only | |
CPTP | |
Human | |
His | |
GLTPD1 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 50% glycerol and 1mM DTT. |