missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IFN-epsilon Protein

Product Code. 18792178
Change view
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18792178 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18792178 Supplier R&D Systems™ Supplier No. 9667ME025/CF
  • ONLINE EXCLUSIVE PRICE
    565.00€
    504.00€ / 25µg
    Save 61.00€
    10% Off
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility.

The Recombinant Human IFN-epsilon Protein is derived from E. coli. The Recombinant Human IFN-epsilon Protein has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q86WN2
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2μm filtered solution in Citric Acid and CHAPS.
Gene ID (Entrez) 338376
Molecular Weight (g/mol) 23 kDa
Name IFNE, IFN-E, IFNE1PRO655, IFNepsilon, IFN-epsilon, IFNT1, Ifn-tau-11, INFE1, interferon epsilon, interferon epsilon 1, Interferon epsilon-1, interferon tau-1, interferon, epsilon, interferon-epsilon, MGC119018, MGC119020
Quantity 25 μg
Source E. coli-derived human IFN-epsilon protein Leu22-Arg208 (Cys175Ser),with an N-terminal Met and a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1μg of the protein by the LAL method.
Conjugate Unconjugated
Species Human
Reconstitution Reconstitute at 250μg/mL in water.
Product Line R&D Systems
Purity or Quality Grade >90%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie Blue Staining.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.