missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IGF-2 / IGF-II Protein
Recombinant Protein
Brand: enQuireBio™ QP5264-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000603.1 | |
IGF-2 / IGF-II Protein | |
Research Use Only | |
IGF2 | |
Recombinant Protein | |
E. coli | |
AYRPSETLCG GELVDTLQFV CGDRGFYFSR PASRVSRRSR GIVEECCFRS CDLALLETYC ATPAKSE | |
Greater than 95% as determined by SDS-PAGE. |
3481 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 15 ng/mL in the Mouse FDC-P1 cell proliferation assay. The bioactivity acceptance criteria is set at 6.7 x 10^4 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |