missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-10 Protein

Product Code. 18737356
Change view
Click to view available options
Quantity:
1 mg
10 μg
250 μg
50 μg
Unit Size:
10µg
1mg
250µg
50µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18737356 10 μg 10µg
18747306 1 mg 1mg
18778256 50 μg 50µg
18717756 250 μg 250µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18737356 Supplier R&D Systems™ Supplier No. 1064ILB010

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Measured in a cell proliferation assay using MC/9-2 mouse mast cells. Thompson-Snipes, L. et al. (1991) J. Exp. Med. 173:507. The ED50 for this effect is 0.075-0.750 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P22301.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3586
Molecular Weight (g/mol) M.W.-observed: 18 kDa, under reducing conditions., M.W.-theroretical: 19 kDa
Quantity 10 μg
Source E. coli-derived human IL-10 protein Ser19-Asn178, with a N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias CSIF, CSIFMGC126450, Cytokine synthesis inhibitory factor, GVHDS, IL10, IL10A, IL-10 mgC126451, interleukin 10, interleukin-10, TGIF
Conjugate Unconjugated
Reconstitution Reconstitute at 100-500 μg/mL in PBS.
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein IL-10
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.