missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL17D Protein

A cDNA sequence encoding the IL17D was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10724-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15914978

  • 4090.00€ / 1mg
Estimated Shipment: 28-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

53342
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
Immobilized rHuIL-17D binds to rHuIL-17BR with EC50 less than 2microg/ml.
Human
Untagged
Lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA.
IL17D Protein
Research Use Only
IL17D
Recombinant Protein
E. coli
APRAGRRPAR PRGCADRPEE LLEQLYGRLA AGVLSAFHHT LQLGPREQAR NASCPAGGRP ADRRFRPPTN LRSVSPWAYR ISYDPARYPR YLPEAYCLCR GCLTGLFGEE DVRFRSAPVY MPTVVLRRTP ACAGGRSVYT EAYVTIPVGC TCVPEPEKDA DSINSSIDKQ GAKLLLGPND APAGP
Greater than 97.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions