missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MYL5 Protein
A cDNA sequence encoding the MYL5 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12770-20ug
Additional Details : Weight : 0.01000kg
Specifications
MYL5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMDQNRD GFIDKEDLKD TYASLGKTNV KDDELDAMLK EASGPINFTM FLNLFGEKLS GTDAEETILN AFKMLDPDGK GKINKEYIKR LLMSQADKMT AEEVDQMFQF ASIDVAGNLD YKALSYVITH GEEKEE | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
MYL5 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 30% glycerol and 0.1M NaCl. |