missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NDUFAF2 Protein
A cDNA sequence encoding the NDUFAF2 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12814-1mg
Additional Details : Weight : 0.01000kg
Specifications
91942 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGWSQDLFRA LWRSLSREVK EHVGTDQFGN KYYYIPQYKN WRGQTIREKR IVEAANKKEV DYEAGDIPTE WEAWIRRTRK TPPTMEEILK NEKHREEIKI KSQDFYEKEK LLSKETSEEL LPPPVQTQIK GHASAPYFGK EEPSVAPSST GKTFQPGSWM PRDGKSHNQ | |
Greater than 85.0% as determined by SDS-PAGE. |
NDUFAF2 Protein | |
Research Use Only | |
NDUFAF2 | |
Human | |
His | |
NDUFAF2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 20% glycerol and 200mM NaCl. |