missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human NDUFB9 Protein

A cDNA sequence encoding the NDUFB9 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP12817-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15991639

  • 212.00€ / 10µg
Estimated Shipment: 27-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

4715
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMAFLASG PYLTHQQKVL RLYKRALRHL ESWCVQRDKY RYFACLMRAR FEEHKNEKDM AKATQLLKEA EEEFWYRQHP QPYIFPDSPG GTSYERYDCY KVPEWCLDDW HPSEKAMYPD YFAKREQWKK LRRESWEREV KQLQEETPPG GPLTEALPPA RKEGDLPPLW WYIVTRPRER PM
Greater than 90.0% as determined by SDS-PAGE.
NDUFB9 Protein
Research Use Only
NDUFB9
Human
His
The NDUFB9 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M Urea and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions