missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDGF-AB Protein
Recombinant Protein
Brand: enQuireBio™ QP5201-1mg
Additional Details : Weight : 0.01000kg
Specifications
PDGF-AB Protein | |
Research Use Only | |
PDGFA/B | |
Recombinant Protein | |
E. coli | |
Alphachain :MSIEEAVPA VCKTRTVIYE IPRSQVDPTS ANFLIWPPCV EVKRCTGCCN TSSVKCQPSR VHHRSVKVAK VEYVRKKPKL KEVQVRLEEH LECACATTSL NPDYREEDTG RPRESGKKRK RKRLKPT Bet achain:MSL GSLTIAEPAM IAECKTRTEV FEISRRLIDR TNANFLVWPP CVEVQRCSGC CNNRNVQCRP TQVQLRPVQV RKIEIVRKKP IFKKATVTLE DHLACKCETV AAARPVT | |
Greater than 95% as determined by SDS-PAGE. |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 20 ng/mL in the BALB/c 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 5.0 x 10^4 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |