missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Peptidyl-prolyl cis-trans isomerase FKBP3 Protein
A DNA sequence encoding the Homo sapiens (Human) Peptidyl-prolyl cis-trans isomerase FKBP3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6049-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002004.1 | |
Peptidyl-prolyl cis-trans isomerase FKBP3 Protein | |
Research Use Only | |
FKBP3 | |
Recombinant Protein | |
E. coli | |
AAAVPQRAWT VEQLRSEQLP KKDIIKFLQE HGSDSFLAEH KLLGNIKNVA KTANKDHLVT AYNHLFETKR FKGTESISKV SEQVKNVKLN EDKPKETKSE ETLDEGPPKY TKSVLKKGDK TNFPKKGDVV HCWYTGTLQD GTVFDTNIQT SAKKKKNAKP LSFKVGVGKV IRGWDEALLT MSKGEKARLE IEPEWAYGKK GQPDAKIPPN AKLTFEVELV DID | |
Greater than 90% as determined by SDS-PAGE. |
2287 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |