missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Peptidyl-prolyl cis-trans isomerase FKBP3 Protein

A DNA sequence encoding the Homo sapiens (Human) Peptidyl-prolyl cis-trans isomerase FKBP3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6049-ec-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15929619

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.


Description

Description

Specifications

Specifications

NP_002004.1
Peptidyl-prolyl cis-trans isomerase FKBP3 Protein
Research Use Only
FKBP3
Recombinant Protein
E. coli
AAAVPQRAWT VEQLRSEQLP KKDIIKFLQE HGSDSFLAEH KLLGNIKNVA KTANKDHLVT AYNHLFETKR FKGTESISKV SEQVKNVKLN EDKPKETKSE ETLDEGPPKY TKSVLKKGDK TNFPKKGDVV HCWYTGTLQD GTVFDTNIQT SAKKKKNAKP LSFKVGVGKV IRGWDEALLT MSKGEKARLE IEPEWAYGKK GQPDAKIPPN AKLTFEVELV DID
Greater than 90% as determined by SDS-PAGE.
2287
50 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions