missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PIH1D2 Protein

Product Code. 15972359
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 μg
Unit Size:
100µg
10µg
2µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15972359 100 μg 100µg
15992359 2 μg 2µg
15982359 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15972359 Supplier enQuireBio™ Supplier No. QP13053100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the PIH1D2 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 120379
Name PIH1D2 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Gene Symbol PIH1D2
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGSSHHHHHH SSGLVPRGSH MGSMETSSKG LLTQVTQFWN LLDDLAQSDP EGYEKFIQQQ LKEGKQLCAA PEPQLCLQTR ILKPKEKILF INLCQWTRIP APQSTTHPVP LTVGKPEDTT EISDAYTVID VAYNPDVLHA AEKDQVKKNQ LIQMAMKCIE EKFQFTLSHS YHITKFRIKG SIQRMKQNLM GIQTDSIDLR EKMRRELTLG QIRSSTMSNP DHFPQLLLPK DQVSGKAVCL IEEISSTEIQ VEMKMPAYEL KIVHDHSEKP LKIELKVELP GINSVSLCDL SVSEDDLLIE VSEKYRLHLN LPKLIDTEMT TAKFIKEKST LIITMPLV
Buffer PIH1D2 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT.
Purity or Quality Grade Greater than 85.0% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.