missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A denatured recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 26-456 of Human PLA1A The Recombinant Human PLA1A Protein is derived from E. coli. The Recombinant Human PLA1A Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Concentration | 1mg/mL |
| For Use With (Application) | SDS-PAGE |
| Formulation | 20mM Tris-HCl buffer (pH 8.0) containing 0.4M UREA, 10% glycerol |
| Gene ID (Entrez) | 51365 |
| Molecular Weight (g/mol) | TMW: 49.5kDa |
| Quantity | 0.5 mg |
| Storage Requirements | Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles. |
| Gene Alias | EC 3.1.1, EC 3.1.1.-, NMD, phosphatidylserine-specific phospholipase A1alpha, phospholipase A1 member A, PS-PLA1, PSPLA1Phosphatidylserine-specific phospholipase A1 |
| Common Name | PLA1A |
| Conjugate | Unconjugated |
| Show More |
For research use only and is not approved for use in humans or in clinical diagnosis
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?