missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
An un-tagged recombinant protein corresponding to the amino acids1-133 of PLA2G16/HRASLS3 The Recombinant Human PLA2G16/HRASLS3 Protein is derived from E. coli. The Recombinant Human PLA2G16/HRASLS3 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | NP_001121675 |
| For Use With (Application) | ELISA, SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) |
| Gene ID (Entrez) | 1145 |
| Molecular Weight (g/mol) | 14.9kDa |
| Name | Nicotinic Acetylcholine Receptor epsilon Protein |
| Purification Method | Protein |
| Immunogen | MRAPIPEPKP GDLIEIFRPF YRHWAIYVGD GYVVHLAPPS EVAGAGAASV MSALTDKAIV KKELLYDVAG SDKYQVNNKH DDKYSPLPCS KIIQRAEELV GQEVLYKLTS ENCEHFVNEL RYGVARSDQV RDV |
| Storage Requirements | −80°C. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?