missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Plexin B1 (Sema domain) His Protein

Product Code. 18759446
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18759446 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18759446 Supplier R&D Systems™ Supplier No. 10803PB050
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Recombinant Protein

Measured by its binding ability in a functional ELISA. When Recombinant Human Plexin B1 His-tag is coated at 1.00 μg/mL (100 μL/well), Recombinant Human Semaphorin 4D Fc Chimera (Catalog # 7470-S4 ) binds with an ED50 of 80.0-480 ng/mL.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O43157.3
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS.
Gene ID (Entrez) 5364
Molecular Weight (g/mol) M.W.-observed: 60-68 kDa, under reducing conditions, M.W.-theroretical: 56 kDa
Quantity 50 μg
Source Chinese Hamster Ovary cell line, CHO-derived human Plexin B1 protein Leu20-Gln535, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU / 1 μg of the protein by the LAL method.
Gene Alias plexin B1, PLXN5, PLXNB1, SEP, Transmembrane Protein SEP
Conjugate Unconjugated
Reconstitution Reconstitute at 1.00 mg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Plexin B1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.