missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-250 of Human PSMA8 The Recombinant Human PSMA8 Protein is derived from E. coli. The Recombinant Human PSMA8 Protein has been validated for the following applications: SDS-Page.
Specifications
Specifications
| Accession Number | Q8TAA3 |
| For Use With (Application) | SDS-PAGE |
| Formulation | Liquid. In 20mM Tris-HCl buffer (pH 8.0) containing 1mM DTT, 10% glycerol, 0.1M NaCl |
| Gene ID (Entrez) | 143471 |
| Molecular Weight (g/mol) | 30kDa |
| Name | PSMA8 Protein |
| Purification Method | SDS-PAGE |
| Storage Requirements | 4°C short term, −20°C long term. Avoid freeze-thaw cycles. |
| Cross Reactivity | Human |
| Purity or Quality Grade | >90% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?